Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380349.1 | NP_001368448.1 | 2 - 3 | 317 | 62 | |
NM_068936.5 | NP_501337.1 | 4 - 5 | 588 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |