Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabs-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307243.2 | NP_001294172.1 | 5 - 6 | 1078 | 149 | |
NM_068951.4 | NP_501352.1 | 5 - 6 | 1080 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |