Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgrs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068974.8 | NP_501375.2 | 5 - 6 | 1610 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |