Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ocr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313034.3 | NP_001299963.1 | 7 - 8 | 1198 | 155 | |
NM_068979.5 | NP_501380.1 | 12 - 13 | 2521 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |