Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ocr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313034.3 | NP_001299963.1 | 2 - 3 | 280 | 161 | |
NM_068979.5 | NP_501380.1 | 7 - 8 | 1603 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |