Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068998.10 | NP_501399.1 | 3 - 4 | 2380 | 61 | |
GenBank mRNA | AB055110.1 | - | 3 - 4 | 2402 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |