Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068998.10 | NP_501399.1 | 4 - 5 | 2550 | 102 | |
GenBank mRNA | AB055110.1 | - | 4 - 5 | 2572 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |