Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380358.1 | NP_001368459.1 | 3 - 4 | 551 | 153 | |
NM_069017.8 | NP_501418.1 | 4 - 5 | 722 | 153 | ||
GenBank mRNA | AY147933.1 | - | 4 - 5 | 752 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |