Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069046.6 | NP_501447.1 | 4 - 5 | 463 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |