Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307101.3 | NP_001294030.1 | 6 - 7 | 815 | 72 | |
NM_001380373.1 | NP_001368453.1 | 5 - 6 | 794 | 72 | ||
NM_069058.9 | NP_501459.1 | 5 - 6 | 803 | 72 | ||
GenBank mRNA | AF105010.1 | - | 5 - 6 | 803 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |