Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069074.7 | NP_501475.1 | 8 - 9 | 2918 | 78 | |
GenBank mRNA | AF163018.1 | - | 8 - 9 | 2903 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |