Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356929.4 | NP_001343640.1 | 8 - 9 | 1780 | 81 | |
NM_001356930.4 | NP_001343703.1 | 8 | 1573 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |