Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028199.5 | NP_001023370.1 | 4 - 5 | 482 | 81 | |
NM_001028200.6 | NP_001023371.1 | 5 - 6 | 693 | 81 | ||
NM_001047522.4 | NP_001040987.1 | 5 - 6 | 699 | 81 | ||
NM_001380387.1 | NP_001367079.1 | 5 - 6 | 693 | 81 | ||
NM_069133.5 | NP_501534.2 | 5 - 6 | 693 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |