Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129334.4 | NP_001122806.1 | 2 - 3 | 284 | 73 | |
NM_001268454.3 | NP_001255383.1 | 1 - 2 | 227 | 73 | ||
NM_069217.8 | NP_501618.2 | 2 - 3 | 284 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |