Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069269.5 | NP_501670.2 | 11 - 12 | 2093 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |