Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307043.3 | NP_001293972.1 | 2 - 3 | 196 | 74 | |
NM_069303.5 | NP_501704.1 | 3 - 4 | 358 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |