Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038346.6 | NP_001033435.1 | 3 - 4 | 485 | 104 | |
NM_001038347.3 | NP_001033436.1 | 1 - 2 | 162 | 104 | ||
NM_001268473.4 | NP_001255402.1 | 3 - 4 | 465 | 104 | ||
NM_001380414.1 | NP_001368126.1 | 2 - 3 | 366 | 104 | ||
NM_001380415.1 | NP_001368482.1 | 3 - 4 | 447 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |