Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038346.6 | NP_001033435.1 | 6 - 7 | 1114 | 68 | |
NM_001038347.3 | NP_001033436.1 | 4 - 5 | 791 | 68 | ||
NM_001380414.1 | NP_001368126.1 | 5 - 6 | 995 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |