Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268483.3 | NP_001255412.1 | 3 - 4 | 551 | 115 | |
NM_001268484.3 | NP_001255413.1 | 4 - 5 | 1041 | 115 | ||
NM_001383168.1 | 4 - 5 | 1024 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |