Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028074.5 | NP_001023245.1 | 2 - 3 | 223 | 67 | |
NM_069315.6 | NP_501716.2 | 2 - 3 | 176 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |