Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047489.4 | NP_001040954.1 | 1 - 2 | 260 | 85 | |
NM_001383169.1 | 1 - 2 | 262 | 85 | |||
GenBank mRNA | AF332211.1 | - | 1 - 2 | 260 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |