Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047489.4 | NP_001040954.1 | 6 - 7 | 1127 | 73 | |
NM_001383169.1 | 6 - 7 | 1002 | 73 | |||
GenBank mRNA | AF332211.1 | - | 6 - 7 | 1127 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |