Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268494.2 | NP_001255423.1 | 2 - 3 | 282 | 108 | |
GenBank mRNA | AF078796.1 | - | 2 - 3 | 240 | 108 | |
AF114440.1 | - | 2 - 3 | 282 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |