Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047534.5 | NP_001040999.2 | 7 - 8 | 1183 | 91 | |
NM_001268500.2 | NP_001255429.1 | 7 - 8 | 1200 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |