Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383170.1 | 3 - 4 | 1243 | 61 | ||
NM_069369.5 | NP_501770.1 | 4 - 5 | 578 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |