Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dur-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268504.3 | NP_001255433.1 | 4 - 5 | 306 | 125 | |
NM_001268505.3 | NP_001255434.1 | 5 - 6 | 372 | 125 | ||
NM_001268506.3 | NP_001255435.1 | 5 - 6 | 375 | 125 | ||
NM_001268507.3 | NP_001255436.1 | 7 - 8 | 1209 | 125 | ||
NM_069385.6 | NP_501786.2 | 5 - 6 | 1087 | 125 | ||
NM_069386.8 | NP_501787.3 | 6 - 7 | 1140 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |