Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069449.7 | NP_501850.1 | 4 - 5 | 643 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |