Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129314.4 | NP_001122786.1 | 11 - 12 | 3392 | 114 | |
NM_069463.6 | NP_501864.2 | 11 - 12 | 3396 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |