Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069537.7 | NP_501938.1 | 9 - 10 | 2537 | 133 | |
GenBank mRNA | AF038576.1 | - | 9 - 10 | 2558 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |