Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tax-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026121.2 | NP_001021292.1 | 10 - 11 | 1493 | 89 | |
NM_001083189.5 | NP_001076658.2 | 10 - 11 | 1493 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |