Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028110.4 | NP_001023281.1 | 11 - 12 | 9870 | 73 | |
NM_001307078.4 | NP_001294007.1 | 11 - 12 | 9841 | 73 | ||
NM_001307079.3 | NP_001294008.1 | 12 - 13 | 9891 | 73 | ||
GenBank mRNA | AB001074.1 | - | 11 - 12 | 9864 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |