Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: miz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268583.2 | NP_001255512.1 | 9 - 10 | 1440 | 65 | |
NM_001268584.2 | NP_001255513.1 | 5 - 6 | 918 | 65 | ||
NM_001307057.4 | NP_001293986.1 | 9 - 10 | 1442 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |