Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268594.3 | NP_001255523.2 | 2 - 3 | 324 | 64 | |
NM_001268595.3 | NP_001255524.1 | 1 - 2 | 67 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |