Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069717.7 | NP_502118.4 | 13 - 14 | 2322 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |