Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069725.5 | NP_502126.1 | 3 - 4 | 536 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |