Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028249.6 | NP_001023420.2 | 1 - 2 | 61 | 56 | |
NM_001028250.5 | NP_001023421.2 | 1 - 2 | 61 | 56 | ||
NM_001307424.2 | NP_001294353.1 | 1 - 2 | 62 | 56 | ||
NM_001307425.1 | NP_001294354.1 | 1 - 2 | 61 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |