Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028249.6 | NP_001023420.2 | 4 - 5 | 677 | 87 | |
NM_001028250.5 | NP_001023421.2 | 4 - 5 | 677 | 87 | ||
NM_001307424.2 | NP_001294353.1 | 4 - 5 | 678 | 87 | ||
NM_001307425.1 | NP_001294354.1 | 4 - 5 | 677 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |