Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028249.6 | NP_001023420.2 | 9 - 10 | 1685 | 88 | |
NM_001028250.5 | NP_001023421.2 | 9 - 10 | 1685 | 88 | ||
NM_001380011.1 | NP_001368238.1 | 5 - 6 | 953 | 88 | ||
NM_001380012.1 | NP_001368239.1 | 5 - 6 | 953 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |