Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067570.6 | NP_499971.2 | 8 - 9 | 1040 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |