Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: immp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380028.1 | NP_001368250.1 | 2 - 3 | 278 | 119 | |
NM_067621.4 | NP_500022.1 | 3 - 4 | 385 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |