Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067672.5 | NP_500073.3 | 1 - 2 | 77 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |