Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067685.5 | NP_500086.1 | 6 - 7 | 1465 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |