Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dsl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380053.1 | NP_001368285.1 | 2 - 3 | 217 | 77 | |
NM_067707.4 | NP_500108.1 | 3 - 4 | 474 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |