Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067738.8 | NP_500139.1 | 5 - 6 | 795 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |