Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cng-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307651.3 | NP_001294580.1 | 2 - 3 | 412 | 61 | |
NM_001356909.3 | NP_001343690.1 | 12 - 13 | 2287 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |