Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xbx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067764.4 | NP_500165.1 | 2 - 3 | 607 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |