Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067770.5 | NP_500171.2 | 6 - 7 | 708 | 131 | |
GenBank mRNA | AY652946.1 | - | 6 - 7 | 701 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |