Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028336.4 | NP_001023507.1 | 2 - 3 | 243 | 88 | |
NM_001383059.1 | 2 - 3 | 227 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |