Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067880.6 | NP_500281.1 | 1 - 2 | 70 | 81 | |
GenBank mRNA | AF020555.1 | - | 1 - 2 | 70 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |