Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067880.6 | NP_500281.1 | 2 - 3 | 164 | 114 | |
GenBank mRNA | AF020555.1 | - | 2 - 3 | 164 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |